EMAIL THIS PAGE TO A FRIEND
T0703 Sigma-Aldrich

D-Talitol

≥98%

  • CAS Number 643-03-8

  • Empirical Formula (Hill Notation) C6H14O6

  • Molecular Weight 182.17

  •  MDL number MFCD00191491

Purchase

Properties

assay   ≥98%
storage temp.   2-8°C
SMILES string   OC[C@@H](O)[C@@H](O)[C@@H](O)[C@H](O)CO

Description

General description

A rare sugar alcohol (polyol).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

X3375 Xylitol, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?