EMAIL THIS PAGE TO A FRIEND
T1136 Sigma-Aldrich

Triphenylphosphine

Synonym: Phosphorustriphenyl

  • CAS Number 603-35-0

  • Linear Formula (C6H5)3P

  • Molecular Weight 262.29

  •  Beilstein/REAXYS Number 610776

  •  EC Number 210-036-0

  •  MDL number MFCD00003043

Purchase

Properties

vapor density   9 (vs air)
vapor pressure   5 mmHg ( 20 °C)
InChI Key   RIOQSEWOXXDEQQ-UHFFFAOYSA-N
bp   377 °C(lit.)
mp   79-81 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Nervous system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SZ3500000
Flash Point(F) 
356 °F
Flash Point(C) 
180 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T84409 Triphenylphosphine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?