EMAIL THIS PAGE TO A FRIEND
T1263 Sigma-Aldrich

Tetrathiafulvalene

~97%

Synonym: Δ2,2′-Bi-1,3-dithiole, TTF

  • CAS Number 31366-25-3

  • Empirical Formula (Hill Notation) C6H4S4

  • Molecular Weight 204.36

  •  Beilstein/REAXYS Number 1617956

  •  EC Number 250-593-7

  •  MDL number MFCD00005492

Purchase

Properties

assay   ~97%
mp   116-119 °C (lit.)
storage temp.   2-8°C
SMILES string   S1C=CS\C1=C2/SC=CS2
InChI   1S/C6H4S4/c1-2-8-5(7-1)6-9-3-4-10-6/h1-4H
InChI key   FHCPAXDKURNIOZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

183180 Tetrathiafulvalene, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?