• USA Home
  • T1267 - Trimethylsilyl-L-(+)-rhamnose

EMAIL THIS PAGE TO A FRIEND
T1267 Sigma-Aldrich

Trimethylsilyl-L-(+)-rhamnose

liquid, Mixed anomers

  • CAS Number 108392-01-4

  • Empirical Formula (Hill Notation) C18H44O5Si4

  • Molecular Weight 452.88

  •  PubChem Substance ID 24899951

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
form   liquid
color   clear colorless
storage temp.   2-8°C
SMILES string   C[C@@H]1OC(O[Si](C)(C)C)[C@H](O[Si](C)(C)C)[C@H](O[Si](C)(C)C)[C@H]1O[Si](C)(C)C
InChI   1S/C18H44O5Si4/c1-14-15(20-24(2,3)4)16(21-25(5,6)7)17(22-26(8,9)10)18(19-14)23-27(11,12)13/h14-18H,1-13H3/t14-,15-,16+,17+,18?/m0/s1
InChI key   QQOFWFOBJUGLLJ-ATICWTEVSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?