• USA Home
  • T1379 - DL-Tryptophan octyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
T1379 Sigma-Aldrich

DL-Tryptophan octyl ester hydrochloride

  • CAS Number 6278-90-6

  • Empirical Formula (Hill Notation) C19H28N2O2 · HCl

  • Molecular Weight 352.90

  •  MDL number MFCD00050620

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?