• USA Home
  • T1427 - Triprolidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
T1427 Sigma-Aldrich

Triprolidine hydrochloride

Sigma Reference Standard

Synonym: (E)-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride, trans-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride

  • CAS Number 6138-79-0

  • Empirical Formula (Hill Notation) C19H22N2 · HCl

  • Molecular Weight 314.85

  •  MDL number MFCD00039044

  •  PubChem Substance ID 329826672

Purchase

Properties

grade   Sigma Reference Standard
packaging   vial of 500 mg
solubility   H2O: moderately soluble
  ethanol: moderately soluble
  methanol: moderately soluble
SMILES string   CC(C=C1)=CC=C1/C(C2=NC=CC=C2)=C\CN3CCCC3.Cl
InChI   1S/C19H22N2.ClH/c1-16-7-9-17(10-8-16)18(19-6-2-3-12-20-19)11-15-21-13-4-5-14-21;/h2-3,6-12H,4-5,13-15H2,1H3;1H/b18-11+;
InChI key   WYUYEJNGHIOFOC-NWBUNABESA-N
Gene Information   human ... HRH1(3269)

Description

Application

Triprolidine hydrochloride has been used as a histamine H1 ligand to assess its affinity for [125I]-iodophenpropit binding sites. Furthermore, triprolidine has also been used to evaluate quantitative structure-retention relationships of antihistamine drugs chromatographed on α1-acid glycoprotein columns.

Biochem/physiol Actions

H1 histamine receptor antagonist.

Packaging

Supplied in amber screw-cap vials

Preparation Note

Triprolidine hydrochloride is moderately soluble in methanol, ethanol and water.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
UT7658000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?