• USA Home
  • T1884 - 8-Thiomethyladenosine 3′:5′-cyclic monophosphate

EMAIL THIS PAGE TO A FRIEND
T1884 Sigma-Aldrich

8-Thiomethyladenosine 3′:5′-cyclic monophosphate

powder

  • CAS Number 61866-10-2

  • Empirical Formula (Hill Notation) C11H14N5O6PS

  • Molecular Weight 375.30

Purchase

Properties

form   powder
color   white
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?