EMAIL THIS PAGE TO A FRIEND
T2017 Sigma-Aldrich

Toluene-d8

  • CAS Number 2037-26-5

  • Linear Formula C6D5CD3

  • Molecular Weight 100.19

  •  Beilstein/REAXYS Number 1909300

  •  EC Number 218-009-5

  •  MDL number MFCD00044638

Purchase

Properties

InChI Key   YXFVVABEGXRONW-JGUCLWPXSA-N
isotopic purity   100 atom % D
packaging   pkg of 10 0.5-mL ampules
refractive index   n20/D 1.494 (lit.)
bp   110 °C(lit.)
density   0.943 g/mL at 25 °C (lit.)
mass shift   M+8
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN1294 - class 3 - PG 2 - Toluene
WGK Germany 
2
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

233382 Toluene-d8, "100%", 99.96 atom % D

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?