EMAIL THIS PAGE TO A FRIEND
T2536 Sigma-Aldrich

Lead(II) tartrate

≥98%

Synonym: L(+)Tartaric acid lead(II) salt, L-2,3-Dihydroxybutanedioic acid lead(II) salt

  • CAS Number 815-84-9

  • Empirical Formula (Hill Notation) C4H4O6Pb

  • Molecular Weight 355.27

  •  Beilstein/REAXYS Number 6120250

  •  EC Number 212-426-6

  •  MDL number MFCD00043282

Purchase

Properties

assay   ≥98%
SMILES string   OC1C(O)C(=O)O[PbH2]OC1=O
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9/PG 3
WGK Germany 
3
RTECS 
OG4800000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?