• USA Home
  • T2785 - Trizma® citrate dibasic solution

EMAIL THIS PAGE TO A FRIEND
T2785 Sigma-Aldrich

Trizma® citrate dibasic solution

1 M

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions, Buffers A to Z More...
Quality Level   200
form   solution
concentration   1 M
pH   5.0-5.2 (0.05 M)
SMILES string   NC(CO)(CO)CO.NC(CO)(CO)CO.OC(=O)CC(O)(CC(O)=O)C(O)=O
InChI   1S/C6H8O7.2C4H11NO3/c7-3(8)1-6(13,5(11)12)2-4(9)10;2*5-4(1-6,2-7)3-8/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*6-8H,1-3,5H2
InChI key   XDJZTRQRZFHVTE-UHFFFAOYSA-N

Description

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Buffers - From Bnech to Bulk

pHast Pack
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?