EMAIL THIS PAGE TO A FRIEND
T3025 Sigma-Aldrich

Thr-Asp

  • CAS Number 108320-97-4

  • Empirical Formula (Hill Notation) C8H14N2O6

  • Molecular Weight 234.21

  •  MDL number MFCD00057849

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(O)C(N)C(=O)NC(CC(O)=O)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?