EMAIL THIS PAGE TO A FRIEND
T3527 Sigma-Aldrich

Melissic acid

~99% (GC)

Synonym: Triacontanoic acid

  • CAS Number 506-50-3

  • Linear Formula CH3(CH2)28COOH

  • Molecular Weight 452.80

  •  Beilstein/REAXYS Number 1803689

  •  EC Number 208-042-3

  •  MDL number MFCD00002813

  •  PubChem Substance ID 24900051

Purchase

Properties

assay   ~99% (GC)
mp   92-94 °C (lit.)
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
InChI   1S/C30H60O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30(31)32/h2-29H2,1H3,(H,31,32)
InChI key   VHOCUJPBKOZGJD-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

134805 Melissic acid, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?