• USA Home
  • T3533 - L-Tyrosine-[ring-3,5-3H]

EMAIL THIS PAGE TO A FRIEND
T3533 Sigma-Aldrich

L-Tyrosine-[ring-3,5-3H]

aqueous solution (containing 2% ethanol)

  • CAS Number 7234-03-9

  • Linear Formula 4-HO-3,5-T2-C6H2CH2CH[NH2]CO2H

  • Molecular Weight 181.19

  •  MDL number MFCD00135819

  •  PubChem Substance ID 24900052

Purchase

Properties

form   aqueous solution (containing 2% ethanol)
extent of labeling   40-60 Ci/mmol
storage temp.   2-8°C
SMILES string   NC(Cc1ccc(O)cc1)C(O)=O

Description

Packaging

serum bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?