• USA Home
  • T3663 - D-(+)-Trehalose dihydrate

EMAIL THIS PAGE TO A FRIEND
T3663 Sigma-Aldrich

D-(+)-Trehalose dihydrate

BioReagent, from Saccharomyces cerevisiae, plant cell culture tested

Synonym: α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside

  • CAS Number 6138-23-4

  • Empirical Formula (Hill Notation) C12H22O11 · 2H2O

  • Molecular Weight 378.33

  •  Beilstein/REAXYS Number 5322018

  •  EC Number 202-739-6

  •  MDL number MFCD00071594

  •  PubChem Substance ID 24900159

Purchase

Properties

biological source   Saccharomyces cerevisiae
InChI Key   DPVHGFAJLZWDOC-PVXXTIHASA-N
product line   BioReagent
suitability   plant cell culture tested
Featured Industry   Agriculture

Description

Application

Trehalose is used in cryobiology to improve the viability and recovery of plant cells exposed to dehydration and freezing/thawing.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Please Enter a Lot Number

Certificate of Origin

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?