• USA Home
  • T3883 - Thymidylyl(3′→5′)-2′-deoxyguanosine ammonium salt

EMAIL THIS PAGE TO A FRIEND
T3883 Sigma-Aldrich

Thymidylyl(3′→5′)-2′-deoxyguanosine ammonium salt

~95%

  • CAS Number 108321-00-2

  • Linear Formula C20H26N7O11P · NH3

  • Molecular Weight 588.47

Purchase

Properties

assay   ~95%
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?