• USA Home
  • T3911 - 2′-Deoxyadenosine 5′-p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
T3911 Sigma-Aldrich

2′-Deoxyadenosine 5′-p-toluenesulfonate

≥85% (HPLC)

Synonym: 5′-O-Tosyl-2′-deoxyadenosine

  • CAS Number 6698-29-9

  • Empirical Formula (Hill Notation) C17H19N5O5S

  • Molecular Weight 405.43

Purchase

Properties

assay   ≥85% (HPLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?