• USA Home
  • T4137 - Glycerol trihexanoate

EMAIL THIS PAGE TO A FRIEND
T4137 Sigma-Aldrich

Glycerol trihexanoate

Grade II-S, ≥90%

Synonym: 1,2,3-Tricaproylglycerol, Glycerol tricapronate, Tricaproin, Trihexanoin

  • CAS Number 621-70-5

  • Linear Formula [CH3(CH2)4COOCH2]2CHOCO(CH2)4CH3

  • Molecular Weight 386.52

  •  Beilstein/REAXYS Number 1806732

  •  EC Number 210-701-5

  •  MDL number MFCD00042905

Purchase

Properties

InChI Key   MAYCICSNZYXLHB-UHFFFAOYSA-N
type   Grade II-S
assay   ≥90%
density   0.98 g/mL (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
MB2700000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T0888 Glycerol trihexanoate, ~99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?