• USA Home
  • T4258 - Trizma® phosphate dibasic

EMAIL THIS PAGE TO A FRIEND
T4258 Sigma-Aldrich

Trizma® phosphate dibasic

reagent grade

Synonym: Di(tris[hydroxymethyl]aminomethane) phosphate

Purchase

Properties

grade   reagent grade
SMILES string   OP(O)(O)=O.NC(CO)(CO)CO.NC(CO)(CO)CO
InChI   1S/2C4H11NO3.H3O4P/c2*5-4(1-6,2-7)3-8;1-5(2,3)4/h2*6-8H,1-3,5H2;(H3,1,2,3,4)
InChI key   MYASHAYBKIUKNJ-UHFFFAOYSA-N

Description

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?