• USA Home
  • T4524 - Tunicamycin B complex

EMAIL THIS PAGE TO A FRIEND
T4524 Sigma

Tunicamycin B complex

~95%

  • CAS Number 66081-36-5

  • Empirical Formula (Hill Notation) C39H64N4O16

  • Molecular Weight 844.94

Purchase

Properties

assay   ~95%
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?