EMAIL THIS PAGE TO A FRIEND
T4568 Sigma-Aldrich

Tyrphostin AG 528

~90%

Synonym: N-(3′,4′-Dihydroxybenzylidenecyanoacetyl)indoline, Tyrphostin B66

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme,
InChI Key   MYKCTDWWIWGLHW-NTEUORMPSA-N
assay   ~90%
potency   IC50 (2.1 micromolar, HER1-2 kinase activity)
  IC50 (4.9 micromolar for EGFK kinase activity)
solubility   DMSO: soluble 10 mg/mL
storage temp.   −20°C
Gene Information   human ... EGFR(1956), ERBB2(2064)

Description

Biochem/physiol Actions

EGFR protein tyrosine kinase inhibitor.

Preparation Note

Tyrphostin AG 528 is soluble in DMSO at 10 mg/ml.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?