• USA Home
  • T4569 - Temechine hydrobromide

EMAIL THIS PAGE TO A FRIEND
T4569 Sigma

Temechine hydrobromide

≥98% (TLC), powder

Synonym: 2,2,6,6-tetramethylquinicludine hydrobromide

  • CAS Number 30015-57-7

  • Empirical Formula (Hill Notation) C11H21N · HBr

  • Molecular Weight 248.20

  •  MDL number MFCD00243113

  •  PubChem Substance ID 329826796

Purchase

Description

Biochem/physiol Actions

Neuronal nicotinic acetylcholine receptor antagonist; potent ganglioblocker.

Preparation Note

Temechine hydrobromide is soluble in water.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VD5859700

Documents

Certificate of Analysis


Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?