• USA Home
  • T4649 - Tetrabutylammonium tetraphenylborate

EMAIL THIS PAGE TO A FRIEND
T4649 Sigma-Aldrich

Tetrabutylammonium tetraphenylborate

Synonym: Tetraphenylboron tetrabutylammonium

  • CAS Number 15522-59-5

  • Linear Formula (CH3CH2CH2CH2)4N[B(C6H5)4]

  • Molecular Weight 561.69

  •  Beilstein/REAXYS Number 3857215

  •  EC Number 239-562-9

  •  MDL number MFCD00011749

Purchase

Properties

mp   230-236 °C (lit.)
SMILES string   CCCC[N+](CCCC)(CCCC)CCCC.c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4
InChI   1S/C24H20B.C16H36N/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h1-20H;5-16H2,1-4H3/q-1;+1
InChI key   ZHCCBGAUZWZGQV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

281034 Tetrabutylammonium tetraphenylborate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?