• USA Home
  • T5020 - 1,2,3,4-Tetra-O-acetyl-α-L(−)-fucose

EMAIL THIS PAGE TO A FRIEND
T5020 Sigma-Aldrich

1,2,3,4-Tetra-O-acetyl-α-L(−)-fucose

Synonym: Tetra-O-acetyl-α-L(−)fucopyranose

Purchase

Properties

InChI Key   QZQMGQQOGJIDKJ-CSHNMWLISA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?