• USA Home
  • T5894 - Tetra-N-acetylchitotetraose

EMAIL THIS PAGE TO A FRIEND
T5894 Sigma-Aldrich

Tetra-N-acetylchitotetraose

~75% (HPLC)

Synonym: NI, NII, NIII, NIV-Tetraacetylchitotetraose

  • CAS Number 2706-65-2

  • Empirical Formula (Hill Notation) C32H54N4O21

  • Molecular Weight 830.79

  •  MDL number MFCD00136030

Purchase

Properties

assay   ~75% (HPLC)
storage temp.   −20°C
SMILES string   CC(=O)NC(C=O)C(O)C(OC1OC(CO)C(OC2OC(CO)C(OC3OC(CO)C(O)C(O)C3NC(C)=O)C(O)C2NC(C)=O)C(O)C1NC(C)=O)C(O)CO
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?