• USA Home
  • T6153 - 4-Aminophenylacetyl-Tuftsin

EMAIL THIS PAGE TO A FRIEND
T6153 Sigma-Aldrich

4-Aminophenylacetyl-Tuftsin

≥97% (HPLC)

Synonym: p-Aminophenylacetyl-Thr-Lys-Pro-Arg

  • CAS Number 63147-94-4

  • Empirical Formula (Hill Notation) C29H47N9O7

  • Molecular Weight 633.74

  •  MDL number MFCD00133732

  •  PubChem Substance ID 24900355

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
composition   Peptide content, ~60%
storage temp.   −20°C
SMILES string   CC(O)C(NC(=O)Cc1ccc(N)cc1)C(=O)NC(CCCCN)C(=O)N2CCCC2C(=O)NC(CCCNC(N)=N)C(O)=O
InChI   1S/C29H47N9O7/c1-17(39)24(37-23(40)16-18-9-11-19(31)12-10-18)26(42)35-20(6-2-3-13-30)27(43)38-15-5-8-22(38)25(41)36-21(28(44)45)7-4-14-34-29(32)33/h9-12,17,20-22,24,39H,2-8,13-16,30-31H2,1H3,(H,35,42)(H,36,41)(H,37,40)(H,44,45)(H4,32,33,34)
InChI key   LWGJHKZQCNVQPO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?