EMAIL THIS PAGE TO A FRIEND
T6193 Sigma-Aldrich

Tyrphostin AG 1112

≥98%, solid

  • CAS Number 153150-84-6

  • Empirical Formula (Hill Notation) C15H10N6

  • Molecular Weight 274.28

  •  PubChem Substance ID 329826843

Purchase

Description

Biochem/physiol Actions

Tyrphostin AG 1112 is a Bcr-Abl kinase blocker that can activate the terminal differentiation of K562 cells and the purging of Ph+ cells. AG 1112 can also interact with anti-Fas receptor antibody to stimulate apoptosis1.

Casein kinase II inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

This compound is featured on the Abl page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Tyrphostin AG 1112 is soluble in DMSO and is insoluble in ethanol.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Phosphorylation is a ubiquitous cellular regulatory mechanism that mediates signal transduction pathways, which carry signals from the cell surface to the nucleus or cytoplasm, by altering the activi...
Keywords: Apoptosis, Bioactive small molecules, Cancer, Diseases, Inflammation, Metabolism, Neurodegenerative Diseases, Phosphorylations, Transduction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?