• USA Home
  • T6533 - Tetraheptylammonium bromide

EMAIL THIS PAGE TO A FRIEND
T6533 Sigma

Tetraheptylammonium bromide

cationic

  • CAS Number 4368-51-8

  • Linear Formula [CH3(CH2)6]4N(Br)

  • Molecular Weight 490.69

  •  Beilstein Registry Number 3763391

  •  EC Number 224-459-3

  •  MDL number MFCD00011861

  •  PubChem Substance ID 24900384

Purchase

Properties

InChI Key   YQIVQBMEBZGFBY-UHFFFAOYSA-M
mp   87-89 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

87301 Tetraheptylammonium bromide, ≥99.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?