EMAIL THIS PAGE TO A FRIEND
T6925 Sigma-Aldrich

Triphenylsilanethiol

98% (GC)

Synonym: Mercaptotriphenylsilane

  • CAS Number 14606-42-9

  • Linear Formula (C6H5)3SiSH

  • Molecular Weight 292.47

  •  Beilstein/REAXYS Number 2941369

  •  MDL number MFCD00192551

Purchase

Properties

InChI Key   MQDRKELSDPESDZ-UHFFFAOYSA-N
assay   98% (GC)
mp   101-104 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

422150 Triphenylsilanethiol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?