• USA Home
  • T7033 - Tetrakis(decyl)ammonium bromide

EMAIL THIS PAGE TO A FRIEND
T7033 Sigma-Aldrich

Tetrakis(decyl)ammonium bromide

>99% (titration)

Synonym: Tetra-decylammonium bromide

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
assay   >99% (titration)
mol wt   659.01 g/mol
SMILES string   [Br-].CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC
InChI   1S/C40H84N.BrH/c1-5-9-13-17-21-25-29-33-37-41(38-34-30-26-22-18-14-10-6-2,39-35-31-27-23-19-15-11-7-3)40-36-32-28-24-20-16-12-8-4;/h5-40H2,1-4H3;1H/q+1;/p-1
InChI key   AHNISXOXSNAHBZ-UHFFFAOYSA-M
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

87580 Tetrakis(decyl)ammonium bromide, ≥99.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?