• USA Home
  • T7266 - Thymidylyl(3′→5′)-2′-deoxyadenosine ammonium salt

EMAIL THIS PAGE TO A FRIEND
T7266 Sigma-Aldrich

Thymidylyl(3′→5′)-2′-deoxyadenosine ammonium salt

  • CAS Number 61845-39-4

  • Empirical Formula (Hill Notation) C20H24N7O10PNH3

  • Molecular Weight 570.45

  •  MDL number MFCD00058630

  •  PubChem Substance ID 24900441

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
storage temp.   −20°C
SMILES string   N.CC1=CN(C2CC(OP(O)(=O)OCC3OC(CC3O)n4cnc5c(N)ncnc45)C(CO)O2)C(=O)NC1=O
InChI   1S/C20H26N7O10P.H3N/c1-9-4-26(20(31)25-19(9)30)15-3-11(12(5-28)35-15)37-38(32,33)34-6-13-10(29)2-14(36-13)27-8-24-16-17(21)22-7-23-18(16)27;/h4,7-8,10-15,28-29H,2-3,5-6H2,1H3,(H,32,33)(H2,21,22,23)(H,25,30,31);1H3
InChI key   DHMLNDOINODMAA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?