• USA Home
  • T8050 - (3aS,4R,6R,7aS)-Tetrahydro-4-hydroxymethyl-6-methoxy-4H-furo[3,2-c]pyran-2(3H)-one

EMAIL THIS PAGE TO A FRIEND
T8050 Sigma-Aldrich

(3aS,4R,6R,7aS)-Tetrahydro-4-hydroxymethyl-6-methoxy-4H-furo[3,2-c]pyran-2(3H)-one

≥98%

Synonym: Methyl 2-deoxy-β-D-alloside 3,4-carbonate

  • CAS Number 62210-98-4

  • Empirical Formula (Hill Notation) C9H14O5

  • Molecular Weight 202.20

  •  PubChem Substance ID 24900498

Purchase

Properties

assay   ≥98%
storage temp.   −20°C

Description

Biochem/physiol Actions

Precursor in synthesis of thromboxane derivatives.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?