EMAIL THIS PAGE TO A FRIEND
T8646 Sigma-Aldrich

L-(−)-Talose

Purchase

Properties

SMILES string   OC[C@H](O)[C@@H](O)[C@@H](O)[C@@H](O)C=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?