• USA Home
  • T8750 - DL-Threonine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
T8750 Sigma-Aldrich

DL-Threonine methyl ester hydrochloride

  • CAS Number 62076-66-8

  • Empirical Formula (Hill Notation) C5H11NO3 · HCl

  • Molecular Weight 169.61

  •  MDL number MFCD00070400

  •  PubChem Substance ID 24900553

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
storage temp.   −20°C
SMILES string   Cl.COC(=O)C(N)C(C)O
InChI   1S/C5H11NO3.ClH/c1-3(7)4(6)5(8)9-2;/h3-4,7H,6H2,1-2H3;1H
InChI key   OZSJLLVVZFTDEY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?