EMAIL THIS PAGE TO A FRIEND
T8766 Sigma

p-Toluidine

powder

Synonym: 4-Aminotoluene, 4-Methylaniline

  • CAS Number 106-49-0

  • Linear Formula CH3C6H4NH2

  • Molecular Weight 107.15

  •  Beilstein Registry Number 471281

  •  EC Number 203-403-1

  •  MDL number MFCD00007906

Purchase

Properties

vapor density   3.9 (vs air)
InChI Key   RZXMPPFPUUCRFN-UHFFFAOYSA-N
form   powder
autoignition temp.   899 °F
expl. lim.   6.6 %
color   tan to brown
bp   200 °C(lit.)
mp   41-46 °C (lit.)
density   0.973 g/mL at 25 °C(lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 3451 6.1 / PGII
WGK Germany 
3
RTECS 
XU3150000
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

461121 p-Toluidine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?