• USA Home
  • T8903 - Trimethylsilyl-D-(−)-fructose

EMAIL THIS PAGE TO A FRIEND
T8903 Sigma-Aldrich

Trimethylsilyl-D-(−)-fructose

≥95%, liquid

  • CAS Number 53538-03-7

  • Empirical Formula (Hill Notation) C21H52O6Si5

  • Molecular Weight 541.06

  •  PubChem Substance ID 24900570

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   PLNWQGWZBNJIQM-CGXNFDGLSA-N
assay   ≥95%
form   liquid
color   clear colorless
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?