EMAIL THIS PAGE TO A FRIEND
T9389 Sigma-Aldrich

Methyl tricosanoate

90-95%, crystalline

Synonym: Tricosanoic acid methyl ester

  • CAS Number 2433-97-8

  • Linear Formula CH3(CH2)21COOCH3

  • Molecular Weight 368.64

  •  Beilstein Registry Number 1796627

  •  EC Number 219-420-2

  •  MDL number MFCD00009349

Purchase

Properties

InChI Key   VORKGRIRMPBCCZ-UHFFFAOYSA-N
assay   90-95%
form   crystalline
color   light yellow
mp   53-56 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?