• USA Home
  • T9758 - Thymidine 5′-monophosphate

EMAIL THIS PAGE TO A FRIEND
T9758 Sigma-Aldrich

Thymidine 5′-monophosphate

98-100%

Synonym: TMP, Thymidylic acid, dTMP

  • CAS Number 365-07-1

  • Empirical Formula (Hill Notation) C10H15N2O8P

  • Molecular Weight 322.21

  •  MDL number MFCD00067349

  •  PubChem Substance ID 24900600

Purchase

Properties

assay   98-100%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XP2090000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?