• USA Home
  • T9885 - 5-Thio-D-glucose 6-phosphate diammonium salt

EMAIL THIS PAGE TO A FRIEND
T9885 Sigma-Aldrich

5-Thio-D-glucose 6-phosphate diammonium salt

95-97%

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
assay   95-97%
storage temp.   −20°C
SMILES string   N.OC1SC(COP(O)(O)=O)C(O)C(O)C1O
InChI   1S/C6H13O8PS.H3N/c7-3-2(1-14-15(11,12)13)16-6(10)5(9)4(3)8;/h2-10H,1H2,(H2,11,12,13);1H3
InChI key   YRYZTTZIXXEIJN-UHFFFAOYSA-N

Description

Biochem/physiol Actions

5-Thio-glucose 6-phosphate is a competitive inhibitor of myo-inositol 1-phophate synthetase.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?