EMAIL THIS PAGE TO A FRIEND
U1508 Sigma-Aldrich

U-75302

oil, ≥95.5% (HPLC)

Synonym: 6-[6-(3-Hydroxy-1E,5Z-undecadienyl)-2-pyridinyl]-1,5-hexanediol

Purchase

Properties

assay   ≥95.5% (HPLC)
form   oil
color   light yellow
solubility   DMSO: 10 mg/mL
shipped in   wet ice
storage temp.   −20°C
SMILES string   CCCCC\C=C/C[C@H](O)\C=C\c1cccc(C[C@H](O)CCCCO)n1
Gene Information   human ... LTB4R(1241)

Description

Biochem/physiol Actions

Selective BLT1 leukotriene B4 receptor antagonist.

Packaging

Product is light and air sensitive and should be packaged and stored under nitrogen with desiccant

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?