• USA Home
  • U4003 - Uridylyl(3′→5′)guanosine ammonium salt

EMAIL THIS PAGE TO A FRIEND
U4003 Sigma-Aldrich

Uridylyl(3′→5′)guanosine ammonium salt

Synonym: UpG

  • CAS Number 108320-85-0

  • Empirical Formula (Hill Notation) C19H24N7O13P · xNH3

  • Molecular Weight 589.41 (free acid basis)

  •  MDL number MFCD00058633

Purchase

Properties

assay   ≥98% (HPLC)
storage temp.   −20°C
SMILES string   N.NC1=NC(=O)c2ncn(C3OC(COP(O)(=O)OC4C(O)C(OC4CO)N5C=CC(=O)NC5=O)C(O)C3O)c2N1
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?