• USA Home
  • U5883 - UCL-1848 trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
U5883 Sigma-Aldrich

UCL-1848 trifluoroacetate salt

≥98% (HPLC), solid

Synonym: 8,14-Diaza-1,7(1,4)-diquinolinacyclotetradecaphane trifluoroacetate salt

  • CAS Number 201147-53-7

  • Empirical Formula (Hill Notation) C32H34F6N4O4 · xC2HF3O2

  • Molecular Weight 652.63 (free base basis)

  •  PubChem Substance ID 329827639

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Ion Channels,
InChI Key   PYNGDRPNACKKCY-WLOGLECFSA-N
assay   ≥98% (HPLC)
form   solid
color   white
solubility   H2O: ~18 mg/mL
storage temp.   −20°C

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

Selective Ca2+-activated, SK potassium channel blocker

Preparation Note

UCL-1848 trifluoroacetate salt is soluble in water at approximately 18 mg/ml.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number

Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?