• USA Home
  • U7252 - Uridylyl(2′→5′)uridine ammonium salt

EMAIL THIS PAGE TO A FRIEND
U7252 Sigma-Aldrich

Uridylyl(2′→5′)uridine ammonium salt

  • CAS Number 108321-54-6

  • Empirical Formula (Hill Notation) C18H22N4O14P · xNH3

  • Molecular Weight 549.36 (free acid basis)

  •  MDL number MFCD00058218

  •  PubChem Substance ID 24900665

Purchase

Properties

storage temp.   −20°C
SMILES string   N.OCC1OC(C(OP(O)(=O)OCC2OC(C(O)C2O)N3C=CC(=O)NC3=O)C1O)N4C=CC(=O)NC4=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?