• USA Home
  • U9376 - Uridine 2′-monophosphate lithium salt

EMAIL THIS PAGE TO A FRIEND
U9376 Sigma-Aldrich

Uridine 2′-monophosphate lithium salt

~98%

  • CAS Number 56070-25-8

  • Empirical Formula (Hill Notation) C9H13N2O9P

  • Molecular Weight 324.18

  •  MDL number MFCD00069754

  •  PubChem Substance ID 24900686

Purchase

Properties

assay   ~98%
storage temp.   −20°C
SMILES string   [Li].OCC1OC(C(OP(O)(O)=O)C1O)N2C=CC(=O)NC2=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?