• USA Home
  • UC134 - (±)-4´-Hydroxynirvanol

EMAIL THIS PAGE TO A FRIEND
UC134 Sigma-Aldrich

(±)-4´-Hydroxynirvanol

Synonym: (±)-5-Ethyl-5-(4-hydroxyphenyl)hydantoin, (±)-5-Ethyl-5-(4-hydroxyphenyl)imidazolinidine-2,4-dione

Purchase

Description

Application

(±)-4′-Hydroxynirvanol can be used for mephenytoin metabolism assays.

Packaging

5, 10 mg in glass bottle

Biochem/physiol Actions

Secondary mephenytoin metabolite; derivative of anticonvulsive agent.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

4′-Hydroxynirvanol can be dissolved in acetone.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
NI9453716

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?