• USA Home
  • UC176 - (R)-(−)-Mephenytoin

EMAIL THIS PAGE TO A FRIEND
UC176 Sigma-Aldrich

(R)-(−)-Mephenytoin

Synonym: (R)-(−)-5-Ethyl-3-methyl-5-phenyl-2,4-imidazolidinedione, (R)-(−)-5-Ethyl-3-methyl-5-phenylhydantoin

  • CAS Number 71140-51-7

  • Empirical Formula (Hill Notation) C12H14N2O2

  • Molecular Weight 218.25

  •  MDL number MFCD00272651

Purchase

Properties

InChI Key   GMHKMTDVRCWUDX-GFCCVEGCSA-N
color   off-white
solubility   acetone: soluble
  chloroform: soluble
  methanol: soluble
storage temp.   2-8°C

Description

Biochem/physiol Actions

CYP2B6 and CYP2C19 substrate; mephenytoin isomer; anticonvulsive; antiepileptic.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

(R)-(−)-Mephenytoin is soluble in chloroform, methanol and acetone.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?