EMAIL THIS PAGE TO A FRIEND
UC428 Sigma-Aldrich

Monoacetyldapsone

Synonym: Mono-N-acetyl-4-4´-sulfonylbisbenzeneamine

  • CAS Number 565-20-8

  • Empirical Formula (Hill Notation) C14H14N2O3S

  • Molecular Weight 290.34

  •  MDL number MFCD00672496

Purchase

Properties

color   white
storage temp.   2-8°C
SMILES string   CC(=O)Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2

Description

Application

Monoacetyldapsone can be used for the chromatographic analysis of dapsone conjugates.

Biochem/physiol Actions

NAT conjugate of dapsone

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. UC428.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AE7020000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?