• USA Home
  • UC456 - Tramadol hydrochloride

EMAIL THIS PAGE TO A FRIEND
UC456 Sigma-Aldrich

Tramadol hydrochloride

≥98% (HPLC), solid

Synonym: (±)-cis-2-(Dimethylaminomethyl)-1-(3-methoxyphenyl)cyclohexanol hydrochloride

Purchase

Properties

Related Categories Approved Therapeutics/Drug Candidates, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Drug and Xenobiotic Metabolism,
assay   ≥98% (HPLC)
form   solid
drug control   USDEA Schedule IV
color   white to off-white
mp   178-181 °C
solubility   H2O: soluble
storage temp.   2-8°C
SMILES string   Cl.COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2CN(C)C
InChI   1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16+;/m1./s1
InChI key   PPKXEPBICJTCRU-XMZRARIVSA-N

Description

Application

Tramadol hydrochloride has been used for pharmacokinetic studies after its oral administration in female goats.

Packaging

5, 10 mg in glass bottle

Biochem/physiol Actions

Opioid analgesic; P450 substrate.

Features and Benefits

This compound was developed by Shionogi. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Tramadol hydrochloride is soluble in water.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN2811 - class 6.1 - PG 3 - MP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?