• USA Home
  • V0875 - DL-Valine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
V0875 Sigma-Aldrich

DL-Valine methyl ester hydrochloride

Purchase

Properties

InChI Key   KUGLDBMQKZTXPW-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?