EMAIL THIS PAGE TO A FRIEND
V109 Sigma-Aldrich

Veratridine

Synonym: 3-Veratroylveracevine, Veracevine veratrate

  • CAS Number 71-62-5

  • Empirical Formula (Hill Notation) C36H51NO11

  • Molecular Weight 673.79

  •  Beilstein/REAXYS Number 78875

  •  EC Number 200-758-4

  •  MDL number MFCD00082515

Purchase

Properties

InChI Key   FVECELJHCSPHKY-FCSYLLAHSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Opens voltage-dependent Na+ channels and prevents their inactivation. This, in turn, opens voltage-activated calcium channels, thus increasing intracellular calcium content and inducing neurotransmitter release. Alkaloid neurotoxin which depolarizes excitable tissue; used to increase membrane sodium permeability. Veratridine is cytotoxic to chromaffin cells in vitro.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1544PSN1 6.1 / PGI
WGK Germany 
3
RTECS 
YX5600000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

V5754 Veratridine, ≥90% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?