• USA Home
  • V1125 - L-Valine t-butyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
V1125 Sigma-Aldrich

L-Valine t-butyl ester hydrochloride

  • CAS Number 13518-40-6

  • Empirical Formula (Hill Notation) C9H19NO2 · HCl

  • Molecular Weight 209.71

  •  MDL number MFCD00034863

  •  PubChem Substance ID 24900709

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
form   crystalline
color   white
SMILES string   Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C
InChI   1S/C9H19NO2.ClH/c1-6(2)7(10)8(11)12-9(3,4)5;/h6-7H,10H2,1-5H3;1H/t7-;/m0./s1
InChI key   AUIVQIHTTVPKFS-FJXQXJEOSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

94660 L-Valine tert-butyl ester hydrochloride, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?